Ethyl-3-hexenoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19734-06
A19734-14
CAAAA19734-06EA
40.45
CAD
CAAAA19734-06
CAAAA19734-14
Ethyl-3-hexenoate 98%
Ethyl-3-hexenoate
Purity: 98%
CAS No.: 2396-83-0
Molecular Formula: CH3CH2CH=CHCH2CO2CH2CH3
Formula Weight: 142.20
Flash Point: 59°(139°F)
Refractive Index: 1.4260
Boiling Point: 63-64°/12mm
UN No.: 3272
MDL No.: MFCD00036524
Formula:
CH₃CH₂CH=CHCH₂CO₂CH₂CH₃ Boiling Pt: 63…64 °C/12 mm Density: 0.89 Flash Pt: 59 °C (139 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00036524 CAS Number: 2396-83-0 EINECS: 219-257-7 UN: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...