Ethyl-3-thiopheneacetate 98%

Supplier: Thermo Scientific Chemicals

A19694-06 A19694-14 A19694-22
CAAAA19694-06EA 76.63 CAD
CAAAA19694-06 CAAAA19694-14 CAAAA19694-22
Ethyl-3-thiopheneacetate 98%
Ethyl-3-thiopheneacetate

Purity: 98% CAS No.: 37784-63-7 Molecular Formula: C8H10O2S Formula Weight: 170.23 Flash Point: 103°(217°F) Refractive Index: 1.5100 Boiling Point: 110-114°/6mm Beilstein Reference: 122555 MDL No.: MFCD00005472

Formula: C₈H₁₀O₂S
Boiling Pt: 110…114 °C/6 mm
Density: 1.137
Flash Pt: 103 °C(217 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005472
CAS Number: 37784-63-7
EINECS: 253-665-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR