2,2,3,3-Tetrafluoropropionic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

A19648-09 A19648-88
CAAAA19648-09EA 316 CAD
CAAAA19648-09 CAAAA19648-88
2,2,3,3-Tetrafluoropropionic acid 97%
2,2,3,3-Tetrafluoropropionic acid

Purity: 97% CAS No.: 756-09-2 Molecular Formula: HCF2CF2CO2H Formula Weight: 146.04 Refractive Index: 1.3210 Boiling Point: 133-134° UN No.: 3265 MDL No.: MFCD00054686

Formula: C₃H₂F₄O₂
MW: 146.04 g/mol
Boiling Pt: 133…134 °C
Density: 1.563
Storage Temperature: Ambient
MDL Number: MFCD00054686
CAS Number: 756-09-2
EINECS: 212-049-7
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR