Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19632-14
A19632-22
CAAAA19632-14EA
94.8
CAD
CAAAA19632-14
CAAAA19632-22
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride 97%
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride
Purity: 97%
CAS No.: 1719-83-1
Molecular Formula: C12H8O6
Formula Weight: 248.19
Melting Point: ca 360°
Beilstein Reference: 294213
MDL No.: MFCD00082228
Formula:
C₁₂H₈O₆ Melting Pt: ca 360 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082228 CAS Number: 1719-83-1 EINECS: 217-009-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...