3-Bromo-2-(bromomethyl)propionic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

A19630-06 A19630-14 A19630-22
CAAAA19630-06EA 124.35 CAD
CAAAA19630-06 CAAAA19630-14 CAAAA19630-22
3-Bromo-2-(bromomethyl)propionic acid 98%
3-Bromo-2-(bromomethyl)propionic acid

Purity: 98% CAS No.: 41459-42-1 Molecular Formula: (BrCH2)2CHCO2H Formula Weight: 245.91 Melting Point: 98-101° UN No.: 3261 MDL No.: MFCD00010643

Formula: C₄H₆Br₂O₂
Melting Pt: 98…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00010643
CAS Number: 41459-42-1
UN: 3261
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR