3-Methylbenzo[b]thiophene-2-acetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19620-02
A19620-88
CAAAA19620-02EA
175.38
CAD
CAAAA19620-02
CAAAA19620-09
CAAAA19620-88
3-Methylbenzo[b]thiophene-2-acetic acid 97%
3-Methylbenzo[b]thiophene-2-acetic acid
Purity: 97%
CAS No.: 1505-52-8
Molecular Formula: C11H10O2S
Formula Weight: 206.25
Melting Point: 148-150°
MDL No.: MFCD00052299
Formula:
C₁₁H₁₀O₂S Melting Pt: 148…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052299 CAS Number: 1505-52-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...