Diethylbenzene (mixture of isomers)

Supplier: Thermo Scientific Chemicals
Danger

A19548-0F A19548-AP
CAAAA19548-0FEA 192.76 CAD
CAAAA19548-0F CAAAA19548-AP
Diethylbenzene (mixture of isomers)
Diethylbenzene (mixture of isomers)

Mixture of isomers


CAS No.: 25340-17-4 Molecular Formula: C10H14 Formula Weight: 134.22 Flash Point: 55°(131°F) Refractive Index: 1.495 Boiling Point: 182-184° UN No.: 2049 MDL No.: MFCD00792900

Formula: C₁₀H₁₄
Boiling Pt: 182…184 °C
Density: 0.87
Flash Pt: 55 °C(131 °F)
Storage Temperature: Ambient
MDL Number: MFCD00792900
CAS Number: 25340-17-4
EINECS: 246-874-9
UN: 2049
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR