Ethyl-2-methyl-4-pentenoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 2-methylpent-4-enoate, 2-Methyl-4-pentenoic Acid Ethyl Ester
A19526-06
A19526-14
A19526-22
CAAAA19526-06EA
42.5
CAD
CAAAA19526-06
CAAAA19526-14
CAAAA19526-22
Ethyl-2-methyl-4-pentenoate 98%
Ethyl-2-methyl-4-pentenoate
Purity: 98%
CAS No.: 53399-81-8
Molecular Formula: C8H14O2
Formula Weight: 142.20
Flash Point: 40°(104°F)
Refractive Index: 1.4160
Boiling Point: 153-155°
Beilstein Reference: 1753406
UN No.: 3272
MDL No.: MFCD00010235
Formula:
C₈H₁₄O₂ MW: 142.20 g/mol Boiling Pt: 153…155 °C Density: 0.880 Flash Pt: 40 °C(104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010235 CAS Number: 53399-81-8 EINECS: 258-520-0 UN: 3272 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...