4-(2-Furyl)-3-buten-2-one cis- and trans-mixture 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19456-06
A19456-14
CAAAAA19456-22EA
493.32
CAD
CAAAAA19456-22
CAAAA19456-06
CAAAA19456-14
4-(2-Furyl)-3-buten-2-one cis- and trans-mixture 98%
4-(2-Furyl)-3-buten-2-one
Purity: 98%
CAS No.: 623-15-4
Molecular Formula: C8H8O2
Formula Weight: 136.15
Flash Point: 103°(217°F)
Melting Point: 33-38°
Boiling Point: 112-115°/10mm
Beilstein Reference: 109696
MDL No.: MFCD00039566
Formula:
C₈H₈O₂ Boiling Pt: 112…115 °C/10 mm Melting Pt: 33…38 °C Flash Pt: 103 °C(217 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039566 CAS Number: 623-15-4 EINECS: 210-774-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...