4-Sulfobenzoic acid monopotassium salt 95%

Supplier: Thermo Scientific Chemicals

A19432-18 A19432-30
CAAAA19432-18EA 83.27 CAD
CAAAA19432-18 CAAAA19432-30
4-Sulfobenzoic acid monopotassium salt 95%
4-Sulphobenzoic acid monopotassium salt

Purity: 95% CAS No.: 5399-63-3 Molecular Formula: C7H5KO5S Formula Weight: 240.28 MDL No.: MFCD00007509

Formula: C₇H₅KO₅S
Storage Temperature: Ambient
MDL Number: MFCD00007509
CAS Number: 5399-63-3
EINECS: 226-435-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR