Ethyl-3-benzoylacrylate predominantly trans 94%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethyl 4-oxo-4-phenylbut-2-enoate

A19426-09 A19426-18
CAAAA19426-09EA 47.08 CAD
CAAAA19426-09 CAAAA19426-18 CAAAA19426-30
Ethyl-3-benzoylacrylate predominantly trans 94%
Ethyl-3-benzoylacrylate

Purity: 94% CAS No.: 17450-56-5 Molecular Formula: C12H12O3 Formula Weight: 204.23 Flash Point: 88°(190°F) Refractive Index: 1.5430 Boiling Point: 184-185°/25mm Beilstein Reference: 2210035 UN No.: 2810 MDL No.: MFCD00011533

Formula: C₁₂H₁₂O₃
MW: 204.23 g/mol
Boiling Pt: 184…185 °C/25 mm
Density: 1.12
Flash Pt: 88 °C(190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011533
CAS Number: 17450-56-5
EINECS: 408-040-4
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR