5,6,7,8,9,10-Hexahydrocyclohept[b]indole 98%

Supplier: Thermo Scientific Chemicals

A19344-03 A19344-06
CAAAA19344-03EA 112.5 CAD
CAAAA19344-03 CAAAA19344-06 CAAAA19344-14
5,6,7,8,9,10-Hexahydrocyclohept[b]indole 98%
5,6,7,8,9,10-Hexahydrocyclohept[b]indole

Purity: 98% CAS No.: 2047-89-4 Molecular Formula: C13H15N Formula Weight: 185.27 Melting Point: 143-146° MDL No.: MFCD00101340

Formula: C₁₃H₁₅N
Melting Pt: 143…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00101340
CAS Number: 2047-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR