Pyridine hydrobromide 98%

Supplier: Thermo Scientific Chemicals
Warning

A19314-0B A19314-18 A19314-30
CAAAA19314-0BEA 255.96 CAD
CAAAA19314-0B CAAAA19314-18 CAAAA19314-30
Pyridine hydrobromide 98%
Pyridine hydrobromide

Purity: 98% CAS No.: 18820-82-1 Molecular Formula: C5H5N.HBr Formula Weight: 160.01 Melting Point: 216-220° MDL No.: MFCD00012801

Formula: C₅H₅N
Melting Pt: 216…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00012801
CAS Number: 18820-82-1
EINECS: 242-600-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR