Dihydrojasmone 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
3-Methyl-2-pentylcyclopent-2-en-1-one
A19298-14
A19298-22
CAAAA19298-14EA
54.51
CAD
CAAAA19298-14
CAAAA19298-22
Dihydrojasmone 97%
Dihydrojasmone
Purity: 97%
CAS No.: 1128-08-1
Molecular Formula: C11H18O
Formula Weight: 166.26
Flash Point: 121°(250°F)
Refractive Index: 1.479
Boiling Point: 120-121°/12mm
Beilstein Reference: 1906471
MDL No.: MFCD00036480
Formula:
C₁₁H₁₈O MW: 166.26 g/mol Boiling Pt: 120…121 °C/12 mm Density: 0.91 Flash Pt: 121 °C(250 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00036480 CAS Number: 1128-08-1 EINECS: 214-434-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...