Sulfamethazine 99%
Supplier: Thermo Scientific Chemicals
A19276-14
A19276-22
CAAAA19276-14EA
40.45
CAD
CAAAA19276-14
CAAAA19276-22
Sulfamethazine 99%
Sulfamethazine
Sulfonamide antibacterial
Purity: 99%
CAS No.: 57-68-1
Molecular Formula: C12H14N4O2S
Formula Weight: 278.34
Melting Point: 198-201°
MDL No.: MFCD00006066
Formula:
C₁₂H₁₄N₄O₂S Melting Pt: 198…201 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006066 CAS Number: 57-68-1 EINECS: 200-346-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...