5-tert-Butyl-2-methyl-3-furoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19258-02
A19258-88
CAAAA19258-02EA
121.82
CAD
CAAAA19258-02
CAAAA19258-88
5-tert-Butyl-2-methyl-3-furoic acid 97%
5-tert-Butyl-2-methyl-3-furoic acid
Purity: 97%
CAS No.: 38422-62-7
Molecular Formula: C10H14O3
Formula Weight: 182.22
Melting Point: 90-94°
MDL No.: MFCD00221068
Formula:
C₁₀H₁₄O₃ Melting Pt: 90…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00221068 CAS Number: 38422-62-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...