3,5-Dihydroxy-4-methylbenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

A19196-03 A19196-06 A19196-14
CAAAA19196-03EA 54.98 CAD
CAAAA19196-03 CAAAA19196-06 CAAAA19196-14
3,5-Dihydroxy-4-methylbenzoic acid 97%
3,5-Dihydroxy-4-methylbenzoic acid

Purity: 97% CAS No.: 28026-96-2 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 266-207° MDL No.: MFCD00002514

Formula: C₈H₈O₄
Melting Pt: 266…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00002514
CAS Number: 28026-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR