O-Methylhydroxylamine hydrochloride 98+%

Supplier: Thermo Scientific Chemicals
Danger

A19188-06 A19188-14
CAAAA19188-06EA 102.7 CAD
CAAAA19188-06 CAAAA19188-14
O-Methylhydroxylamine hydrochloride 98+%
O-Methylhydroxylamine hydrochloride

Fieser: 1,670 11,322 13,177 21,265


Purity: 98+% CAS No.: 593-56-6 Molecular Formula: CH3ONH2.HCl Formula Weight: 83.52 Melting Point: ca 148° dec. Beilstein Reference: 3589723 Merck Reference: 14,5989 UN No.: 3261 MDL No.: MFCD00012951

Formula: CH₆ClNO
Melting Pt: ca 148 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012951
CAS Number: 593-56-6
EINECS: 209-798-7
UN: 3261
ADR: "8",III
Merck Index: 14,5989

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR