Geranylacetone mixture E/Z 97% (Z)-isomer (nerylacetone) ∼ 45%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19184-14
A19184-22
CAAAA19184-14EA
110.28
CAD
CAAAA19184-14
CAAAA19184-22
Geranylacetone mixture E/Z 97% (Z)-isomer (nerylacetone) ∼ 45%
(E/Z)-Geranylacetone
Purity: 97%,
CAS No.: 3796-70-1
Molecular Formula: (CH3)2C=CHCH2CH2C(CH3)=CHCH2CH2C(O)CH3
Formula Weight: 194.32
Storage Temperature: 2°C to 8°C
Flash Point: 110°(230°F)
Refractive Index: 1.467
Boiling Point: 250°
Beilstein Reference: 1722277
MDL No.: MFCD00008910
Formula:
(CH₃)₂C=CHCH₂CH₂C(CH₃)=CHCH₂CH₂C(O)CH₃ Boiling Pt: 250 °C Density: 0.87 Flash Pt: 110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008910 CAS Number: 3796-70-1 EINECS: 211-711-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...