(+)-Nootkatone 98, Crystalline
Supplier: Thermo Scientific Chemicals
Synonyms:
(4R,4aS,6R)-4,4a-Dimethyl-6-(prop-1-en-2-yl)-4,4a,5,6,7,8-hexahydronaphthalen-2(3H)-one
A19166-14
A19166-03
A19166-06
CAAAAA19166-14EA
1564.2
CAD
CAAAAA19166-14
CAAAA19166-03
CAAAA19166-06
(+)-Nootkatone 98, Crystalline
(+)-Nootkatone
Purity: 98+%
CAS No.: 4674-50-4
Molecular Formula: C15H22O
Formula Weight: 218.34
Storage Temperature: 2°C to 8°C
Flash Point: 99°(210°F)
Melting Point: 34-36°
Boiling Point: 125°/0.5mm
Beilstein Reference: 4676969
Optical Rotation: +185° (c=1 in chloroform)
MDL No.: MFCD00036591
Formula:
C₁₅H₂₂O MW: 218.34 g/mol Boiling Pt: 125 °C (0.5 mmHg) Melting Pt: 34…36 °C Density: 0.997 Flash Pt: 99 °C (210 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD04974578 CAS Number: 4674-50-4 EINECS: 225-124-4 |
Specification Test Results
Optical rotation | +185° (c=1 in chloroform) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...