Guanylurea sulfate 97%

Supplier: Thermo Scientific Chemicals
Warning

A19106-09 A19106-18 A19106-30
CAAAA19106-09EA 51.98 CAD
CAAAA19106-09 CAAAA19106-18 CAAAA19106-30
Guanylurea sulfate 97%
Guanylurea sulfate

Purity: 97% CAS No.: 591-01-5 Molecular Formula: C4H12N8O2.H2SO4 Formula Weight: 302.27 Melting Point: ca 195° dec. Beilstein Reference: 6099371 MDL No.: MFCD00035565

Formula: C₂H₈N₄O₅S
Melting Pt: ca 195 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00035565
CAS Number: 591-01-5
EINECS: 209-697-8
Merck Index: 14,3093

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR