5-Acetylindan 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(2,3-Dihydro-1H-inden-5-yl)ethan-1-one, 1-(2,3-Dihydro-1H-inden-5-yl)ethanone

A19102-09 A19102-18 A19102-88
CAAAA19102-09EA 304.94 CAD
CAAAA19102-09 CAAAA19102-18 CAAAA19102-88
5-Acetylindan 97%
5-Acetylindan

Purity: 97% CAS No.: 4228-10-8 Molecular Formula: C11H12O Formula Weight: 160.22 Boiling Point: 85-87°/0.05mm MDL No.: MFCD00021246

Formula: C₁₁H₁₂O
MW: 160.22 g/mol
Boiling Pt: 85…87 °C/0.05 mm
Storage Temperature: Ambient
MDL Number: MFCD00021246
CAS Number: 4228-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR