3-Nonen-2-one 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Non-3-en-2-one

A19084-30 A19084-09 A19084-18
CAAAAA19084-30EA 439.24 CAD
CAAAAA19084-30 CAAAA19084-09 CAAAA19084-18
3-Nonen-2-one 96%
3-Nonen-2-one

Purity: 96% CAS No.: 14309-57-0 Molecular Formula: C9H16O Formula Weight: 140.23 Flash Point: 81°(177°F) Refractive Index: 1.4490 Boiling Point: 85°/12mm Beilstein Reference: 1743558 MDL No.: MFCD00010241

Formula: C₉H₁₆O
MW: 140.23 g/mol
Boiling Pt: 85 °C/12 mm
Density: 0.848
Flash Pt: 81 °C(177 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010241
CAS Number: 14309-57-0
EINECS: 238-248-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR