Boc-ON ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(tert-BOC-Oxyimino)-2-phenylacetonitrile, N-((tert-Butoxycarbonyl)oxy)benzimidoyl cyanide, 2-(Boc-oxyimino)-2-phenylacetonitrile, 2-(tert-Butoxycarbonyloxyimino)-2-phenylacetonitrile, [(cyano-phenyl-methylidene)amino] tert-butyl carbonate
Fieser: 6,91 10,61
Purity: 98+%
CAS No.: 58632-95-4
Molecular Formula: C13H14N2O3
Formula Weight: 246.27
Storage Temperature: 2°C to 8°C
Melting Point: 86-89°
Beilstein Reference: 2217296
UN No.: 3439
MDL No.: MFCD00001863
Formula:
C13H14N2O3 MW: 246.27 g/mol Melting Pt: 86…89 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001863 CAS Number: 58632-95-4 EINECS: 261-370-9 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...