2-Nitrobenzylamine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Nitrophenyl)methanamine hydrochloride

A18900-03 A18900-06
CAAAA18900-03EA 98.91 CAD
CAAAA18900-03 CAAAA18900-06
2-Nitrobenzylamine hydrochloride 98%
2-Nitrobenzylamine hydrochloride

Purity: 98% CAS No.: 24835-08-3 Molecular Formula: C7H8N2O2.HCl Formula Weight: 188.62 Melting Point: 250° dec. MDL No.: MFCD00136280

Formula: C₇H₈N₂O₂·HCl
MW: 188.62 g/mol
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00136280
CAS Number: 24835-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR