Perfluorophenanthrene mixture of isomers
Supplier: Thermo Scientific Chemicals
A18898-14
A18898-22
CAAAA18898-14EA
114.87
CAD
CAAAA18898-14
CAAAA18898-22
Perfluorophenanthrene mixture of isomers
Perfluorophenanthrene
CAS No.: 306-91-2
Molecular Formula: C14F24
Formula Weight: 624.12
Refractive Index: 1.3348
Melting Point: -20°
Boiling Point: 215°
MDL No.: MFCD00042596
Formula:
C₁₄F₂₄ Boiling Pt: 215 °C Melting Pt: –20 °C Density: 2.030 Storage Temperature: Ambient |
MDL Number:
MFCD00042596 CAS Number: 306-91-2 EINECS: 400-470-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...