(4,5-Dimethoxy-2-nitrophenyl)acetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18884-03
A18884-06
CAAAA18884-03EA
115.34
CAD
CAAAA18884-03
CAAAA18884-06
(4,5-Dimethoxy-2-nitrophenyl)acetic acid 97%
(4,5-Dimethoxy-2-nitrophenyl)acetic acid
Purity: 97%
CAS No.: 73357-18-3
Molecular Formula: C10H11NO6
Formula Weight: 241.20
Melting Point: 206-208°
MDL No.: MFCD00118466
Formula:
C₁₀H₁₁NO₆ Melting Pt: 206…208 °C Storage Temperature: Ambient |
MDL Number:
MFCD00118466 CAS Number: 73357-18-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...