(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: ((1,3-Dioxolan-2-yl)methyl)triphenylphosphonium bromide

A18882-09 A18882-18 A18882-30
CAAAA18882-09EA 121.66 CAD
CAAAA18882-09 CAAAA18882-18 CAAAA18882-30
(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide 98%
(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide

Purity: 98% CAS No.: 52509-14-5 Molecular Formula: C22H22BrO2P Formula Weight: 429.30 Storage Temperature: 2°C to 8°C Melting Point: 186-189° MDL No.: MFCD00011966

Formula: C₂₂H₂₂BrO₂P
MW: 429.30 g/mol
Melting Pt: 186…189 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011966
CAS Number: 52509-14-5
EINECS: 257-977-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR