Nerolidol (cis and trans mixture) ≥97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol, 3-Hydroxy-3,7,11-trimethyl-1,6,10-dodecatriene, Nerolidol, 3,7,11-Trimethyldodeca-1,6,10-trien-3-ol

A18812-14 A18812-22
CAAAA18812-14EA 66.52 CAD
CAAAA18812-14 CAAAA18812-22 CAAAA18812-36
Nerolidol (cis and trans mixture) ≥97%
Nerolidol (cis and trans mixture)

Purity: 97+% CAS No.: 7212-44-4 Molecular Formula: C15H26O Formula Weight: 222.37 Flash Point: 96°(204°F) Refractive Index: 1.4790 Boiling Point: 263-266° Beilstein Reference: 1724135 MDL No.: MFCD00008911

Formula: C₁₅H₂₆O
MW: 222.37 g/mol
Boiling Pt: 263…266 °C
Density: 0.876
Flash Pt: 96 °C (204 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008911
CAS Number: 7212-44-4
EINECS: 230-597-5
UN: 3082
ADR: 9 III
Merck Index: 14,6477

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR