trans-2-Hexenyl acetate 98%

Supplier: Thermo Scientific Chemicals
Warning

A18786-18 A18786-30
CAAAA18786-18EA 61.62 CAD
CAAAA18786-18 CAAAA18786-30
trans-2-Hexenyl acetate 98%
trans-2-Hexenyl acetate

Purity: 98% CAS No.: 2497-18-9 Molecular Formula: C8H14O2 Formula Weight: 142.20 Flash Point: 58°(136°F) Refractive Index: 1.4270 Boiling Point: 165-166° Beilstein Reference: 1721851 UN No.: 3272 MDL No.: MFCD00009474

Formula: C₈H₁₄O₂
MW: 142.20 g/mol
Boiling Pt: 165…166 °C
Density: 0.900
Flash Pt: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009474
CAS Number: 2497-18-9
EINECS: 219-680-7
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR