9-Fluoreneacetic acid ≥99.5%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(9H-Fluoren-9-yl)acetic acid

A18772-03 A18772-06
CAAAA18772-03EA 121.82 CAD
CAAAA18772-03 CAAAA18772-06 CAAAA18772-14
9-Fluoreneacetic acid ≥99.5%
9-Fluoreneacetic acid

Purity: 99.5+% CAS No.: 6284-80-6 Molecular Formula: C15H12O2 Formula Weight: 224.26 Melting Point: 133-135° MDL No.: MFCD00013262

Formula: C₁₅H₁₂O₂
MW: 224.26 g/mol
Melting Pt: 133…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00013262
CAS Number: 6284-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR