Ethyl-3-(2-furyl)propionate 98%

Supplier: Thermo Scientific Chemicals

A18732-06 A18732-14
CAAAA18732-06EA 63.52 CAD
CAAAA18732-06 CAAAA18732-14
Ethyl-3-(2-furyl)propionate 98%
Ethyl-3-(2-furyl)propionate

Purity: 98% CAS No.: 10031-90-0 Molecular Formula: C9H12O3 Formula Weight: 168.19 MDL No.: MFCD00036496

Formula: C₉H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00036496
CAS Number: 10031-90-0
EINECS: 233-097-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR