1-(2-Naphthoyl)-3,3,3-trifluoroacetone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18726-06
A18726-14
CAAAA18726-06EA
80.11
CAD
CAAAA18726-06
CAAAA18726-14
1-(2-Naphthoyl)-3,3,3-trifluoroacetone 99%
1-(2-Naphthoyl)-3,3,3-trifluoroacetone
Purity: 99%
CAS No.: 893-33-4
Molecular Formula: C14H9F3O2
Formula Weight: 266.22
Melting Point: 74-76°
MDL No.: MFCD00054501
Formula:
C₁₄H₉F₃O₂ Melting Pt: 74…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00054501 CAS Number: 893-33-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...