Bisabolene mixture of isomers 96% (sum of isomers)

Supplier: Thermo Scientific Chemicals
Warning

A18724-09 A18724-18
CAAAA18724-09EA 80.26 CAD
CAAAA18724-09 CAAAA18724-18
Bisabolene mixture of isomers 96% (sum of isomers)
Bisabolene

CAS No.: 495-62-5 Molecular Formula: C15H24 Formula Weight: 204.36 Refractive Index: 1.4940 MDL No.: MFCD00129080

Formula: C₁₅H₂₄
Density: 0.89
Storage Temperature: Ambient
MDL Number: MFCD00129080
CAS Number: 495-62-5
EINECS: 207-805-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR