N-(3-Acetyl-4-hydroxyphenyl)butyramide 97%
Supplier: Thermo Scientific Chemicals
A18672-0B
A18672-18
CAAAA18672-0BEA
658.86
CAD
CAAAA18672-0B
CAAAA18672-18
CAAAA18672-30
N-(3-Acetyl-4-hydroxyphenyl)butyramide 97%
N-(3-Acetyl-4-hydroxyphenyl)butyramide
Purity: 97%
CAS No.: 40188-45-2
Molecular Formula: C12H15NO3
Formula Weight: 221.26
Melting Point: 124-126°
MDL No.: MFCD00798556
Formula:
C₁₂H₁₅NO₃ Melting Pt: 124…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD00798556 CAS Number: 40188-45-2 EINECS: 254-831-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...