beta-Alanine benzyl ester p-toluenesulfonate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-β-Ala-OBzl·p-tosylate, H-β-Ala-Obzl.TosOH, H-β-Ala-OBzl·TosOH

A18650-09 A18650-18
CAAAA18650-09EA 99.22 CAD
CAAAA18650-09 CAAAA18650-18 CAAAA18650-30
beta-Alanine benzyl ester p-toluenesulfonate 98%
beta-Alanine benzyl ester p-toluenesulfonate

Purity: 98% CAS No.: 27019-47-2 Molecular Formula: C6H5CH2O2CCH2CH2NH2.HO3SC6H4CH3 Formula Weight: 351.42 Storage Temperature: 2°C to 8°C Melting Point: 138-139° MDL No.: MFCD00039061

Formula: C₁₀H₁₃NO₂
MW: 351.42 g/mol
Melting Pt: 138…139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039061
CAS Number: 27019-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR