Sodium-DL-2-hydroxybutyrate ≥97%

Supplier: Thermo Scientific Chemicals

A18636-03 A18636-06 A18636-14
CAAAA18636-03EA 55.46 CAD
CAAAA18636-03 CAAAA18636-06 CAAAA18636-14
Sodium-DL-2-hydroxybutyrate ≥97%
Sodium-DL-2-hydroxybutyrate

Purity: 97+% CAS No.: 5094-24-6 Molecular Formula: C4H7NaO3 Formula Weight: 126.09 Melting Point: 153-157° Beilstein Reference: 4825104 MDL No.: MFCD00004565

Formula: C₄H₈O₃
Melting Pt: 153…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00004565
CAS Number: 5094-24-6
EINECS: 225-812-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR