8-Ethoxyquinoline-5-sulfonic acid hydrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Actinoquinol
A18622-09
A18622-18
CAAAA18622-09EA
71.26
CAD
CAAAA18622-09
CAAAA18622-18
CAAAA18622-30
8-Ethoxyquinoline-5-sulfonic acid hydrate 99%
8-Ethoxyquinoline-5-sulfonic acid hydrate
Purity: 99%
CAS No.: 15301-40-3
Molecular Formula: C11H11NO4S.H2O
Formula Weight: 271.29 (253.28anhy)
Melting Point: ca 296° dec.
Beilstein Reference: 225878
MDL No.: MFCD00065059
Formula:
C₁₁H₁₁NO₄S MW: 253.27 g/mol Melting Pt: ∼ 296 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00065059 CAS Number: 15301-40-3 EINECS: 239-334-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...