Methyl-4-tert-butylbenzoate 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
Methyl 4-(tert-butyl)benzoate
A18619-14
A18619-22
CAAAA18619-14EA
45.98
CAD
CAAAA18619-14
CAAAA18619-22
CAAAA18619-36
Methyl-4-tert-butylbenzoate 98+%
Methyl 4-tert-butylbenzoate
Purity: 98+%
CAS No.: 26537-19-9
Molecular Formula: C12H16O2
Formula Weight: 192.26
Flash Point:
Formula:
C₁₂H₁₆O₂ MW: 192.26 g/mol Boiling Pt: 122…124 °C (10 mmHg) Density: 0.995 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008835 CAS Number: 26537-19-9 EINECS: 247-768-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...