2-Coumaranone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzofuran-2(3H)-one, Benzo[b]furan-2(3H)-one

A18617-03 A18617-06 A18617-14
CAAAA18617-03EA 69.2 CAD
CAAAA18617-03 CAAAA18617-06 CAAAA18617-14
2-Coumaranone 99%
2-Coumaranone

Purity: 99% CAS No.: 553-86-6 Molecular Formula: C8H6O2 Formula Weight: 134.13 Flash Point: 110°(230°F) Odor: Characteristic Boiling Point: 248-250° Beilstein Reference: 114689 MDL No.: MFCD00005856

Formula: C₈H₆O₂
MW: 134.13 g/mol
Boiling Pt: 248…250 °C
Melting Pt: 47…52 °C
Density: 1.22
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005856
CAS Number: 553-86-6
EINECS: 209-052-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR