4-(Phenylazo)diphenylamine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18612-06
A18612-14
A18612-22
CAAAA18612-06EA
64.62
CAD
CAAAA18612-06
CAAAA18612-14
CAAAA18612-22
4-(Phenylazo)diphenylamine 97%
4-(Phenylazo)diphenylamine
Purity: 97%
CAS No.: 101-75-7
Molecular Formula: C18H15N3
Formula Weight: 273.34
Melting Point: 80-86°
MDL No.: MFCD00003023
Formula:
C₁₈H₁₅N₃ Melting Pt: 80…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003023 CAS Number: 101-75-7 EINECS: 202-972-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...