Uridine 5′-monophosphate disodium salt 99%
Supplier: Thermo Scientific Chemicals
A18601-06
A18601-14
A18601-22
CAAAA18601-06EA
123.87
CAD
CAAAA18601-06
CAAAA18601-14
CAAAA18601-22
Uridine 5′-monophosphate disodium salt 99%
Uridine 5′-monophosphate disodium salt
Purity: 99%
CAS No.: 3387-36-8
Molecular Formula: C9H11N2Na2O9P
Formula Weight: 368.15
Storage Temperature: 2°C to 8°C
Melting Point: ca 209° dec.
Beilstein Reference: 3582885
MDL No.: MFCD00006525
Formula:
C9H11N2Na2O9P Melting Pt: ca 209 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00006525 CAS Number: 3387-36-8 EINECS: 222-211-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...