Nitrofurazone ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Nitrofurfural Semicarbazone, 2-((5-Nitrofuran-2-yl)methylene)hydrazinecarboxamide, 5-Nitro-2-Furaldehyde Semicarbazone

A18593-14 A18593-22 A18593-30
CAAAAA18593-14EA 52.61 CAD
CAAAAA18593-14 CAAAAA18593-22 CAAAA18593-30
Nitrofurazone ≥98%
Nitrofurazone

Bactericidal compound used as an antibiotic


Purity: 98+% CAS No.: 59-87-0 Molecular Formula: C6H6N4O4 Formula Weight: 198.14 Melting Point: ca 240° dec. Beilstein Reference: 86403 MDL No.: MFCD00003225

Formula: C₆H₆N₄O₄
MW: 198.14 g/mol
Melting Pt: ca 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003225
CAS Number: 59-87-0
EINECS: 200-443-1
Merck Index: 14,6600

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR