Eosin Y (yellowish)

Supplier: Thermo Scientific Chemicals

Synonyms: Solvent Red 43, Field's Stain B, Eosin 225, 2,4,5,7-Tetrabromofluorescein disodium salt, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Tetrabromofluorescein disodium salt, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Eosin Y disodium salt

A18578-09 A18578-18
CAAAA18578-09EA 49.61 CAD
CAAAA18578-09 CAAAA18578-18
Eosin Y (yellowish)
Eosin Y (yellowish)

Used to stain blood smears to identify malarial parasites


CAS No.: 17372-87-1 MDL No.: MFCD00146926

Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005040
CAS Number: 17372-87-1
Merck Index: 13,03634

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR