Nalpha-Acetyl-D-arginine dihydrate 98%
Supplier: Thermo Scientific Chemicals
A18563-03
A18563-06
CAAAA18563-03EA
70.63
CAD
CAAAA18563-03
CAAAA18563-06
Nalpha-Acetyl-D-arginine dihydrate 98%
Nalpha-Acetyl-D-arginine dihydrate
Purity: 99%
CAS No.: 2389-86-8
Molecular Formula: C8H16N4O3.2H2O
Formula Weight: 252.29 (216.25anhy)
Melting Point: ca 270°
MDL No.: MFCD00209627
Formula:
C₂₇H₃₃P Melting Pt: ∼ 270 °C Storage Temperature: Ambient |
MDL Number:
MFCD00209627 CAS Number: 23897-15-6 EINECS: 219-225-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...