Salbutamol sulfate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(tert-Butylamino)-1-(4-hydroxy-3-hydroxymethylphenyl)ethanol sulfate, Bronchospray, Albuterol hemisulfate, Proventil-HFA, 5-[2-(tert-Butylamino)-1-hydroxyethyl]-2-hydroxybenzyl Alcohol sulfate, Salbutamol hemisulphate, Salbutamol hemisulfate, Albuterol hemissulphate, 2-(tert-Butylamino)-1-(4-hydroxy-3-hydroxymethylphenyl)ethanol sulphate, Albutarol sulphate, 5-[2-(tert-Butylamino)-1-hydroxyethyl]-2-hydroxybenzyl Alcohol sulphate, Albutarol sulfate

A18544-03 A18544-06
CAAAA18544-03EA 80.58 CAD
CAAAA18544-03 CAAAA18544-06 CAAAA18544-14
Salbutamol sulfate 99%
Salbutamol sulphate

A beta-adrenergic agonist


Purity: 99% CAS No.: 51022-70-9 Molecular Formula: C26H42N2O6.H2SO4 Formula Weight: 576.70 MDL No.: MFCD00055200

Formula: (C₁₃H₂₁NO₃)₂.H₂SO₄
Storage Temperature: Ambient
MDL Number: MFCD00055200
CAS Number: 51022-70-9
EINECS: 256-916-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR