Phthalimidoacetaldehyde diethyl acetal 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18491-06
A18491-14
CAAAA18491-06EA
60.51
CAD
CAAAA18491-06
CAAAA18491-14
Phthalimidoacetaldehyde diethyl acetal 99%
Phthalimidoacetaldehyde diethyl acetal
Purity: 99%
CAS No.: 78902-09-7
Molecular Formula: C14H17NO4
Formula Weight: 263.30
Melting Point: 71-74°
Beilstein Reference: 215793
Formula:
C₁₄H₁₇NO₄ Melting Pt: 71…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005901 CAS Number: 78902-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...