Cyclohexylamine hydrobromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18439-22
A18439-36
CAAAA18439-22EA
57.2
CAD
CAAAA18439-22
CAAAA18439-36
Cyclohexylamine hydrobromide 98%
Cyclohexylamine hydrobromide
Purity: 98%
CAS No.: 26227-54-3
Molecular Formula: C6H13N.HBr
Formula Weight: 180.09
Melting Point: 197-201°
Beilstein Reference: 3908098
MDL No.: MFCD00034986
Formula:
C₆H₁₃N·HBr Melting Pt: 197…201 °C Storage Temperature: Ambient |
MDL Number:
MFCD00034986 CAS Number: 26227-54-3 EINECS: 247-526-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...