7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
7-Hydroxy-5-methyl-1,3,4-triazaindolizine, 5-Methyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol, 4-Hydroxy-6-methyl-1,3,3a,7-tetraazaindene
A18438-14
A18438-22
CAAAA18438-14EA
88.8
CAD
CAAAA18438-14
CAAAA18438-22
7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine 98%
7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine
Purity: 98%
CAS No.: 2503-56-2
Molecular Formula: C6H6N4O
Formula Weight: 150.14
Melting Point: 281-285°
Beilstein Reference: 609440
MDL No.: MFCD00005555
Formula:
C₆H₆N₄O MW: 150.14 g/mol Melting Pt: 281…285 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005555 CAS Number: 2503-56-2 EINECS: 219-706-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...