Erbium(III) oxalate decahydrate 99.9%

Supplier: Thermo Scientific Chemicals
Warning

A18420-06 A18420-14 A18420-22
CAAAAA18420-06EA 45.19 CAD
CAAAAA18420-06 CAAAAA18420-14 CAAAAA18420-22
Erbium(III) oxalate decahydrate 99.9%
Erbium (III) oxalate decahydrate

Purity: 99.9% CAS No.: 30618-31-6 Molecular Formula: C6Er2O12.10H2O Formula Weight: 778.73 (598.58anhy) UN No.: 3288 MDL No.: MFCD00150142

Formula: C₆Er₂O₁₂·10H₂O
Density: 2.64
Storage Temperature: Ambient
MDL Number: MFCD00150142
CAS Number: 30618-31-6
EINECS: 212-764-4
UN: 3288
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR